missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylphenol, 99%, ACROS Organics™

Brand:  Acros Organics 131620250

Code : 29

Additional Details : CAS Number : 645-56-7 Weight : 0.07500kg Transport : UN number : 3145 Chem class : 8 Pack group : III


Packaging Glass bottle
Quantity 25g

 View more versions of this product

Product Code. 10423510

  • 98.30 € / 25g


Estimated Shipment
07-08-2018
Add to basket


Specifications

Chemical Identifiers

4-n-Propylphenol
98.5
99%
C9H12O
MFCD00002395
645-56-7
100.0
99%
CH3CH2CH2C6H4OH

Specifications

4-n-Propylphenol
25g
98.5
Authentic
1.5220 to 1.5240
Liquid
99%
CH3CH2CH2C6H4OH
0.9830g/mL
Clear yellow
KLSLBUSXWBJMEC-UHFFFAOYSA-N
CCCC1=CC=C(C=C1)O
136.194
CHEBI:34434
136.19
645-56-7
100.0
Glass bottle
0.983
99%
C9H12O
MFCD00002395
232.0°C
22.0°C
106°C
4-propylphenol
12580
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-n-Propylphenol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS