missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylphenol, 99%, Thermo Scientific Chemicals

Catalog Number p-4228884
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
645-56-7
MFCD00002395
KLSLBUSXWBJMEC-UHFFFAOYSA-N
4-n-propylphenol, p-propylphenol, phenol, 4-propyl, dihydrochavicol, phenol, p-propyl, 1-hydroxy-4-n-propylbenzene, p-n-propylphenol, 4-propyl-phenol, p-hydroxypropylbenzene, hydroxypropylbenzene, p
12580
CHEBI:34434
4-propylphenol
CCCC1=CC=C(C=C1)O
This item is not returnable. View return policy
645-56-7
MFCD00002395
KLSLBUSXWBJMEC-UHFFFAOYSA-N
4-n-propylphenol, p-propylphenol, phenol, 4-propyl, dihydrochavicol, phenol, p-propyl, 1-hydroxy-4-n-propylbenzene, p-n-propylphenol, 4-propyl-phenol, p-hydroxypropylbenzene, hydroxypropylbenzene, p
12580
CHEBI:34434
4-propylphenol
CCCC1=CC=C(C=C1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 22.0°C
Color Yellow
Density 0.9830g/mL
Boiling Point 232.0°C
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Molecular Formula C9H12O
Linear Formula CH3CH2CH2C6H4OH
Refractive Index 1.5220 to 1.5240
Quantity 1 g
Specific Gravity 0.983
Molecular Weight (g/mol) 136.194
Formula Weight 136.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-n-Propylphenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-n-Propylphenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'