missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-2,3-lutidine N-oxide, 97%, ACROS Organics™

Brand:  Acros Organics 441980250

Code : 29

Additional Details : CAS Number : 37699-43-7 Weight : 0.07500kg


Quantity 25g

 View more versions of this product

Product Code. 11956421

  • 187.00 € / 25g

Estimated Shipment
28-06-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

37699-43-7
168.15
CFMTVTYBZMKULI-UHFFFAOYSA-N
148223
CC1=C(C)[N+]([O-])=CC=C1[N+]([O-])=O
C7H8N2O3
MFCD00065172
2,3-dimethyl-4-nitropyridine-n-oxide, 2,3-dimethyl-4-nitropyridine 1-oxide, 2,3-dimethyl-4-nitropyridine n-oxide, 4-nitro-2,3-lutidine-n-oxide, pyridine, 2,3-dimethyl-4-nitro-, 1-oxide, 2,3-dimethyl-4-nitropyridin-1-ium-1-olate, 4-nitro-2,3-lutidine n-oxide, 4-nitro-2,3-dimethylpyridine n-oxide, 2,3-dimethyl-4-nitro-pyridine 1-oxide, 2,3-dimethyl-4-nitro-1-oxido-pyridin-1-ium
2,3-dimethyl-4-nitro-1-oxidopyridin-1-ium

Specifications

4-Nitro-2, 3-lutidine N-oxide
Yellow
25g
96.0
96% min. (HPLC)
MFCD00065172
CFMTVTYBZMKULI-UHFFFAOYSA-N
2,3-dimethyl-4-nitro-1-oxidopyridin-1-ium
148223
Powder or Crystalline Powder
Authentic
94°C to 96°C
37699-43-7
100.0
C7H8N2O3
2,3-dimethyl-4-nitropyridine-n-oxide, 2,3-dimethyl-4-nitropyridine 1-oxide, 2,3-dimethyl-4-nitropyridine n-oxide, 4-nitro-2,3-lutidine-n-oxide, pyridine, 2,3-dimethyl-4-nitro-, 1-oxide, 2,3-dimethyl-4-nitropyridin-1-ium-1-olate, 4-nitro-2,3-lutidine n-oxide, 4-nitro-2,3-dimethylpyridine n-oxide, 2,3-dimethyl-4-nitro-pyridine 1-oxide, 2,3-dimethyl-4-nitro-1-oxido-pyridin-1-ium
CC1=C(C)[N+]([O-])=CC=C1[N+]([O-])=O
168.15
168.15
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Nitro-2,3-lutidine N-oxide

Signal Word
  • Warning

Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS Documents