Learn More
4-Nitrophenyl Disodium Orthophosphate, Pure, Fisher Chemical
Brand: Fisher Chemical N/3390/46
Code : 21
Additional Details : CAS Number : 333338-18-4 Weight : 0.02500kg
Quantity | 25g |
---|
Chemical Identifiers
4264-83-9 | |
263.052 | |
VIYFPAMJCJLZKD-UHFFFAOYSA-L | |
77949 | |
C1=CC(=CC=C1[N+](=O)[O-])OP(=O)([O-])[O-].[Na+].[Na+] |
C6H4NNa2O6P | |
66288 | |
pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system | |
disodium;(4-nitrophenyl) phosphate |
Specifications
4264-83-9 | |
66288 | |
VIYFPAMJCJLZKD-UHFFFAOYSA-L | |
disodium;(4-nitrophenyl) phosphate | |
77949 | |
Powder Solid | |
White | |
25g |
C6H4NNa2O6P | |
pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system | |
C1=CC(=CC=C1[N+](=O)[O-])OP(=O)([O-])[O-].[Na+].[Na+] | |
263.052 | |
371.13g/mol | |
Plastic powder jar | |
300°C |