missing translation for 'onlineSavingsMsg'
Learn More

4-(p-Carboxyphenyl)-1,2,4-triazole, 97%, Thermo Scientific™

Catalog Number p-7077332
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
157069-48-2
C9H7N3O2
189.17
QCNDLFQKKMPZKH-UHFFFAOYSA-N
4-4h-1,2,4-triazol-4-yl benzoic acid, 4-1,2,4-triazol-4-yl benzoic acid, 4-1,2,4 triazol-4-yl-benzoic acid, benzoic acid, 4-4h-1,2,4-triazol-4-yl, 4-4-carboxyphenyl-1,2,4-triazole, 4-4-carboxyphenyl-4h-1,2,4-triazole, 4-4h-1,2,4-triazol-4-yl-benzoic acid
3163581
4-(1,2,4-triazol-4-yl)benzoic acid
C1=CC(=CC=C1C(=O)O)N2C=NN=C2
This item is not returnable. View return policy
157069-48-2
C9H7N3O2
189.17
QCNDLFQKKMPZKH-UHFFFAOYSA-N
4-4h-1,2,4-triazol-4-yl benzoic acid, 4-1,2,4-triazol-4-yl benzoic acid, 4-1,2,4 triazol-4-yl-benzoic acid, benzoic acid, 4-4h-1,2,4-triazol-4-yl, 4-4-carboxyphenyl-1,2,4-triazole, 4-4-carboxyphenyl-4h-1,2,4-triazole, 4-4h-1,2,4-triazol-4-yl-benzoic acid
3163581
4-(1,2,4-triazol-4-yl)benzoic acid
C1=CC(=CC=C1C(=O)O)N2C=NN=C2
Melting Point 330.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 4-(p-Carboxyphenyl)-1,2,4-triazole
compliance-icons
Product Identifier
  • 4-(p-Carboxyphenyl)-1,2,4-triazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'