missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pyrid-4-ylbenzoic acid, 95%, Maybridge™
Brand: Maybridge CC35401CB
Code : 31
Additional Details : Weight : 0.05025kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 250mg |
Chemical Identifiers
4385-76-6 | |
199.209 | |
DZLGZIGLHCRIMF-UHFFFAOYSA-N | |
1520811 | |
C1=CC(=CC=C1C2=CC=NC=C2)C(=O)O |
C12H9NO2 | |
MFCD04114574 | |
4-pyridin-4-yl-benzoic acid, 4-4-pyridyl benzoic acid, 4-pyridin-4-yl benzoic acid, 4-pyrid-4-ylbenzoic acid, 4-4-pyridinyl benzoic acid, 4-4-carboxyphenyl pyridine, 4'-pyridyl-4-benzoic acid, benzoic acid, 4-4-pyridinyl, pubchem22345, acmc-1adxv | |
4-pyridin-4-ylbenzoic acid |
Specifications
4-Pyrid-4-ylbenzoic acid | |
250mg | |
94.5 | |
95% | |
MFCD04114574 | |
DZLGZIGLHCRIMF-UHFFFAOYSA-N | |
4-pyridin-4-ylbenzoic acid | |
1520811 | |
95% |
Amber Glass Bottle | |
4385-76-6 | |
100.0 | |
C12H9NO2 | |
4-pyridin-4-yl-benzoic acid, 4-4-pyridyl benzoic acid, 4-pyridin-4-yl benzoic acid, 4-pyrid-4-ylbenzoic acid, 4-4-pyridinyl benzoic acid, 4-4-carboxyphenyl pyridine, 4'-pyridyl-4-benzoic acid, benzoic acid, 4-4-pyridinyl, pubchem22345, acmc-1adxv | |
C1=CC(=CC=C1C2=CC=NC=C2)C(=O)O | |
199.209 | |
199.21 |