missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)benzylchloride, 97%, ACROS Organics™

Brand:  Acros Organics 345490250

Code : 29

Additional Details : CAS Number : 19692-45-6 Weight : 0.07500kg Transport : UN number : 1760 Chem class : 8 Pack group : II


Packaging Glass Bottle
Quantity 25g

 View more versions of this product

Product Code. 10011541

  • 103.00 € / 25g

Estimated Shipment: 16-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

19692-45-6
182.69
WAXIFMGAKWIFDQ-UHFFFAOYSA-N
88198
CC(C)(C)C1=CC=C(CCl)C=C1
C11H15Cl
MFCD00000918
4-tert-butylbenzyl chloride, 1-tert-butyl-4-chloromethyl benzene, p-tert-butylbenzyl chloride, 4-tert-butyl benzyl chloride, benzene, 1-chloromethyl-4-1,1-dimethylethyl, 4-tert-butyl benzylchloride, p-t-butylbenzyl chloride, 1-chloromethyl-4-tert-butylbenzene, p-tert-butyl-.alpha.-chlorotoluene
1-tert-butyl-4-(chloromethyl)benzene

Specifications

4-(tert-Butyl)benzylchloride
94°C
Glass Bottle
0.94
Colorless to Yellow
97%
96.0
96% min. (GC)
(CH3)3CC6H4CH2Cl
4-tert-butylbenzyl chloride, 1-tert-butyl-4-chloromethyl benzene, p-tert-butylbenzyl chloride, 4-tert-butyl benzyl chloride, benzene, 1-chloromethyl-4-1,1-dimethylethyl, 4-tert-butyl benzylchloride, p-t-butylbenzyl chloride, 1-chloromethyl-4-tert-butylbenzene, p-tert-butyl-.alpha.-chlorotoluene
CC(C)(C)C1=CC=C(CCl)C=C1
182.69
182.69
97%
0.9400g/mL
Authentic
1.5205 to 1.5225
98°C to 103°C (7.0mmHg)
25g
19692-45-6
100.0
C11H15Cl
MFCD00000918
WAXIFMGAKWIFDQ-UHFFFAOYSA-N
1-tert-butyl-4-(chloromethyl)benzene
88198
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-(tert-Butyl)benzylchloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents
Certificates