missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)iodobenzene, 97%, Thermo Scientific Chemicals

Catalog Number 11974721
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
103962-05-6
C7H4F3IO
288.01
MFCD00042411
RTUDBROGOZBBIC-UHFFFAOYSA-N
1-iodo-4-trifluoromethoxy benzene, 4-trifluoromethoxy iodobenzene, 4-iodotrifluoromethoxybenzene, 1-iado-4-trifluoromethoxy benzene, p-trifluoromethoxy iodobenzene, 4-trifluoromethoxy-iodobenzene, 1-iodo-4-trifluoromethoxy-benzene, benzene, 1-iodo-4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-4-iodoanisole, 4-iodo-alpha,alpha,alpha-trifluoromethoxybenzene
2777294
1-iodo-4-(trifluoromethoxy)benzene
FC(F)(F)OC1=CC=C(I)C=C1
This item is not returnable. View return policy
103962-05-6
C7H4F3IO
288.01
MFCD00042411
RTUDBROGOZBBIC-UHFFFAOYSA-N
1-iodo-4-trifluoromethoxy benzene, 4-trifluoromethoxy iodobenzene, 4-iodotrifluoromethoxybenzene, 1-iado-4-trifluoromethoxy benzene, p-trifluoromethoxy iodobenzene, 4-trifluoromethoxy-iodobenzene, 1-iodo-4-trifluoromethoxy-benzene, benzene, 1-iodo-4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-4-iodoanisole, 4-iodo-alpha,alpha,alpha-trifluoromethoxybenzene
2777294
1-iodo-4-(trifluoromethoxy)benzene
FC(F)(F)OC1=CC=C(I)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.8400g/mL
Boiling Point 72.0°C to 74.0°C (15.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula IC6H4OCF3
Refractive Index 1.504
Quantity 5 g
Specific Gravity 1.84
Formula Weight 288
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)iodobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Trifluoromethoxy)iodobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'