missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included

Brand:  Hamilton™ 7650-01

Code : 13

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10070892

  • 112.00 € / Each
Estimated Shipment: 13-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • PTFE coating from point for 0.75 in.
  • For dispensing both liquids and gases
  • Precision-machined polymer plunger tip that creates leak-free seal

Available as replacement needle for installing into standard removable needle (RN) syringe; Needles sold separately

Specifications

Specifications

22 G
1002
2.5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers