Learn More
Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included
Brand:  Hamilton™ 7639-01
Code : A0
Additional Details : Weight : 0.03200kg
 View more versions of this product
Product Code. 10476323
  • 101.00 € / Each
Estimated Shipment: 03-05-2024
to see stock.
Description
Description
  • ideal for dispensing volumes from 0.05 µL up to 5 µL
  • The 7000 series is the only zero dead volume syringe
  • The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle
Specifications
Specifications
26s G
725
250 μL