missing translation for 'onlineSavingsMsg'
Learn More

5-[2-Methyl-3-(trifluoromethyl)-1H-2lambda~5~-pyrazol-5-yl]-2-thiophenecarboxylic acid, 97%, Maybridge™

Brand:  Maybridge MO07338DA

Code : 31

Additional Details : CAS Number : 223499-20-5 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Packaging Amber Glass Bottle
Quantity 1g

 View more versions of this product

Product Code. 10022314

  • 141.77 € / 1g

Estimated Shipment
11-06-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

223499-20-5
276.233
MGYVPTDPOYVOQK-UHFFFAOYSA-N
2775669
CN1C(=CC(=N1)C2=CC=C(S2)C(=O)O)C(F)(F)F
C10H7F3N2O2S
MFCD00206674
5-1-methyl-5-trifluoromethyl pyrazol-3-yl thiophene-2-carboxylic acid, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl thiophene-2-carboxylic acid, 3-5-carboxythien-2-yl-1-methyl-5-trifluoromethyl-1h-pyrazole, 2-thiophenecarboxylicacid, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl-thiophene-2-carboxylic acid, 5-1-methyl-5-trifluoromethyl-1h-pyrazole-3-yl-2-thiophenecarboxylic acid
5-[1-methyl-5-(trifluoromethyl)pyrazol-3-yl]thiophene-2-carboxylic acid

Specifications

5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carboxylic acid
1g
97.0
C10H7F3N2O2S
5-1-methyl-5-trifluoromethyl pyrazol-3-yl thiophene-2-carboxylic acid, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl thiophene-2-carboxylic acid, 3-5-carboxythien-2-yl-1-methyl-5-trifluoromethyl-1h-pyrazole, 2-thiophenecarboxylicacid, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl-thiophene-2-carboxylic acid, 5-1-methyl-5-trifluoromethyl-1h-pyrazole-3-yl-2-thiophenecarboxylic acid
CN1C(=CC(=N1)C2=CC=C(S2)C(=O)O)C(F)(F)F
276.233
276.24
Amber Glass Bottle
223499-20-5
97%
MFCD00206674
MGYVPTDPOYVOQK-UHFFFAOYSA-N
5-[1-methyl-5-(trifluoromethyl)pyrazol-3-yl]thiophene-2-carboxylic acid
2775669
97%
Videos
SDS Documents