missing translation for 'onlineSavingsMsg'
Learn More

5,5-Diphenylhydantoin sodium salt, 99%, ACROS Organics™

Brand:  Acros Organics 436101000

Code : 29

Additional Details : CAS Number : 630-93-3 Weight : 0.15000kg


Packaging Glass bottle
Quantity 100g

 View more versions of this product

Product Code. 11946251

  • 55.96 € / 100g

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

630-93-3
274.26
FJPYVLNWWICYDW-UHFFFAOYSA-M
131674077
[Na+].O=C1NC(C(=O)[N-]1)(C1=CC=CC=C1)C1=CC=CC=C1
C15H11N2NaO2
MFCD00069674,MFCD00005264
Phenytoin sodium, Sodium 5, 5-diphenyl-2, 4-imidazolidinedione
5,5-diphenylimidazolidine-2,4-dione;molecular hydrogen;sodium

Specifications

5, 5-Diphenylhydantoin sodium salt
98.5
99%
MFCD00069674,MFCD00005264
Phenytoin sodium, Sodium 5, 5-diphenyl-2, 4-imidazolidinedione
FJPYVLNWWICYDW-UHFFFAOYSA-M
5,5-diphenylimidazolidine-2,4-dione;molecular hydrogen;sodium
131674077
Crystalline Powder
Authentic
3% Max.
295°C to 298°C
630-93-3
100.0
C15H11N2NaO2
15,7433
Solubility in water: practically insoluble
[Na+].O=C1NC(C(=O)[N-]1)(C1=CC=CC=C1)C1=CC=CC=C1
274.26
274.25
99%
Glass bottle
White
100g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5,5-Diphenylhydantoin sodium salt

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS Documents
Certificates