missing translation for 'onlineSavingsMsg'
Learn More

5,5-Diphenylhydantoin sodium salt, 99%, Thermo Scientific Chemicals

Catalog Number p-4258709
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
630-93-3
C15H11N2NaO2
274.26
MFCD00069674,MFCD00005264
FJPYVLNWWICYDW-UHFFFAOYSA-M
Phenytoin sodium, Sodium 5, 5-diphenyl-2, 4-imidazolidinedione
131674077
[Na+].O=C1NC(C(=O)[N-]1)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
630-93-3
C15H11N2NaO2
274.26
MFCD00069674,MFCD00005264
FJPYVLNWWICYDW-UHFFFAOYSA-M
Phenytoin sodium, Sodium 5, 5-diphenyl-2, 4-imidazolidinedione
131674077
[Na+].O=C1NC(C(=O)[N-]1)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 295°C to 298°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99%
Merck Index 15,7433
Solubility Information Solubility in water: practically insoluble
IUPAC Name sodium 2,5-dioxo-4,4-diphenylimidazolidin-1-ide
Formula Weight 274.25
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 5, 5-Diphenylhydantoin sodium salt
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 5,5-Diphenylhydantoin sodium salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'