missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-7-azaindole, 97%, Thermo Scientific Chemicals

Catalog Number p-4228421
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
183208-35-7
C7H5BrN2
197.04
MFCD06659677
LPTVWZSQAIDCEB-UHFFFAOYSA-N
10307932
5-bromo-1H-pyrrolo[2,3-b]pyridine
BrC1=CN=C2NC=CC2=C1
This item is not returnable. View return policy
183208-35-7
C7H5BrN2
197.04
MFCD06659677
LPTVWZSQAIDCEB-UHFFFAOYSA-N
10307932
5-bromo-1H-pyrrolo[2,3-b]pyridine
BrC1=CN=C2NC=CC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 179.7°C to 180.7°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Synonym 5-bromo-7-azaindole, 5-bromo-1h-pyrrolo 2,3-b pyridine, 1h-pyrrolo 2,3-b pyridine, 5-bromo, 5-bromo azaindole, 3-bromo-7h-pyrrolo 2,3-b pyridine, 5-bromo-1h-pyrrolo 2 , 3-b pyridine, 5-bromopyrrolo 2,3-b pyridine, 5-bromo,7-azaindole
Formula Weight 197.04
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-7-azaindole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Bromo-7-azaindole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'