Learn More
5-Bromoindole-2-carboxylic acid, 98%, ACROS Organics™
Brand: Acros Organics 398650050
Code : 29
Additional Details : CAS Number : 7254-19-5 Weight : 0.05500kg
Packaging | Glass bottle |
---|---|
Quantity | 5g |
Chemical Identifiers
7254-19-5 | |
240.05 | |
YAULOOYNCJDPPU-UHFFFAOYSA-N | |
252137 | |
C1=CC2=C(C=C1Br)C=C(N2)C(=O)O |
C9H6BrNO2 | |
MFCD00022705 | |
5-bromoindole-2-carboxylic acid, 5-bromo-2-indolecarboxylic acid, 1h-indolecarboxylic acid, 5-bromo, 1h-indole-2-carboxylic acid, 5-bromo, pubchem1663, zlchem 1343, acmc-1bb5x, chembl22435, 5-bromo indole carboxylic acid, 5-bromanyl-1h-indole-2-carboxylic acid | |
5-bromo-1H-indole-2-carboxylic acid |
Specifications
5-Bromoindole-2-carboxylic acid | |
5g | |
97.5 | |
97.5% min. (HPLC) | |
MFCD00022705 | |
YAULOOYNCJDPPU-UHFFFAOYSA-N | |
5-bromo-1H-indole-2-carboxylic acid | |
252137 | |
98% |
Glass bottle | |
7254-19-5 | |
100.0 | |
C9H6BrNO2 | |
5-bromoindole-2-carboxylic acid, 5-bromo-2-indolecarboxylic acid, 1h-indolecarboxylic acid, 5-bromo, 1h-indole-2-carboxylic acid, 5-bromo, pubchem1663, zlchem 1343, acmc-1bb5x, chembl22435, 5-bromo indole carboxylic acid, 5-bromanyl-1h-indole-2-carboxylic acid | |
C1=CC2=C(C=C1Br)C=C(N2)C(=O)O | |
240.05 | |
240.05 |
Safety and Handling
- 5-Bromoindole-2-carboxylic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.