missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Cyano-2-(trifluoromethoxy)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H61177.03

Code : NEW

Additional Details : CAS Number : 1072946-64-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15476326

  • 752.00 € / 1g


Estimated Shipment date
from Supplier 22-05-2019
Add to basket

Specifications

Chemical Identifiers

1072946-64-5
230.937
5-cyano-2-trifluoromethoxy phenylboronic acid, 5-cyano-2-trifluoromethoxy phenyl boronic acid, 5-cyano-2-trifluoromethoxy phenyl boronic acid, acmc-2098sk
[5-cyano-2-(trifluoromethoxy)phenyl]boronic acid
C8H5BF3NO3
SZPKBNMEIASWNB-UHFFFAOYSA-N
46738976
B(C1=C(C=CC(=C1)C#N)OC(F)(F)F)(O)O

Specifications

5-Cyano-2-(trifluoromethoxy)benzeneboronic acid
1072946-64-5
5-cyano-2-trifluoromethoxy phenylboronic acid, 5-cyano-2-trifluoromethoxy phenyl boronic acid, 5-cyano-2-trifluoromethoxy phenyl boronic acid, acmc-2098sk
B(C1=C(C=CC(=C1)C#N)OC(F)(F)F)(O)O
230.937
230.94
97%
1g
C8H5BF3NO3
SZPKBNMEIASWNB-UHFFFAOYSA-N
[5-cyano-2-(trifluoromethoxy)phenyl]boronic acid
46738976
Solid
Videos
SDS