missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroquinazolin-4-ol, 97%, Maybridge™

Brand:  Maybridge BTB04061DA

Code : 31

Additional Details : CAS Number : 436-72-6 Weight : 0.05100kg

Quantity 1g

 View more versions of this product

Product Code. 10019091

  • 16.88 € / 1g

Estimated Shipment
22-02-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

436-72-6
164.139
UXEZULVIMJVIFB-UHFFFAOYSA-N
2737331
C1=CC2=C(C(=C1)F)C(=O)N=CN2
C8H5FN2O
MFCD00173674
5-fluoro-4-hydroxyquinazoline, 5-fluoroquinazolin-4-ol, 5-fluoroquinazolin-4 1h-one, 5-fluoroquinazolin-4 3h-one, 5-fluoro-4-quinazolone, 5-fluoro-3h-quinazolin-4-one, 4 1h-quinazolinone, 5-fluoro, pubchem6307, 5-fluoroquinazolin-4-one, acmc-209jv3
5-fluoro-1H-quinazolin-4-one

Specifications

5-Fluoroquinazolin-4-ol
1g
96.5
97%
MFCD00173674
UXEZULVIMJVIFB-UHFFFAOYSA-N
5-fluoro-1H-quinazolin-4-one
2737331
97%
Amber Glass Bottle
436-72-6
100.0
C8H5FN2O
5-fluoro-4-hydroxyquinazoline, 5-fluoroquinazolin-4-ol, 5-fluoroquinazolin-4 1h-one, 5-fluoroquinazolin-4 3h-one, 5-fluoro-4-quinazolone, 5-fluoro-3h-quinazolin-4-one, 4 1h-quinazolinone, 5-fluoro, pubchem6307, 5-fluoroquinazolin-4-one, acmc-209jv3
C1=CC2=C(C(=C1)F)C(=O)N=CN2
164.139
164.14
Videos
SDS Documents