missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-methoxypyridine, 97%, ACROS Organics™

Brand:  Acros Organics 438930050

Code : 29

Additional Details : CAS Number : 13472-61-2 Weight : 0.05500kg


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 11946391

  • 378.00 € / 5g

In stock, ready to ship: 10

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

13472-61-2
235.03
2-methoxy-5-iodopyridine, pyridine, 5-iodo-2-methoxy, pyridine,5-iodo-2-methoxy, pubchem6606, acmc-1c1li, 5-iodanyl-2-methoxy-pyridine
5-iodo-2-methoxypyridine
C6H6INO
NTXRNCUPGYOZCN-UHFFFAOYSA-N
23423786
COC1=NC=C(C=C1)I

Specifications

5-Iodo-2-methoxypyridine
Glass bottle
Colorless to Yellow
13472-61-2
100.0
C6H6INO
NTXRNCUPGYOZCN-UHFFFAOYSA-N
5-iodo-2-methoxypyridine
23423786
Liquid
Authentic
106°C to 108°C (30.0 Torr)
5g
96.0
96% min. (GC)
2-methoxy-5-iodopyridine, pyridine, 5-iodo-2-methoxy, pyridine,5-iodo-2-methoxy, pubchem6606, acmc-1c1li, 5-iodanyl-2-methoxy-pyridine
COC1=NC=C(C=C1)I
235.03
235.03
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Iodo-2-methoxypyridine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS Documents