missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxybenzimidazole, 97%, Thermo Scientific™

Catalog Number 10776275
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 4887-80-3
Melting Point 121°C to 126°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C8H8N2O
Quantity 5 g
Synonym 5-methoxybenzimidazole, 5-methoxy-1h-benzo d imidazole, 5-methoxy-1h-benzimidazole, 1h-benzimidazole, 5-methoxy, 6-methoxy-1h-benzoimidazole, 5-methoxy-1h-1,3-benzodiazole, 5ob, 5-methoxy-3h-1,3-benzodiazole, 5-methoxybenzimazole, pubchem7662
InChI Key ILMHAGCURJPNRZ-UHFFFAOYSA-N
SMILES COC1=CC2=C(C=C1)N=CN2
IUPAC Name 6-methoxy-1H-benzimidazole
Molecular Weight (g/mol) 148.16
PubChem CID 78598
Formula Weight 148.16
Percent Purity 97%
Chemical Name or Material 5-Methoxybenzimidazole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methoxybenzimidazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'