missing translation for 'onlineSavingsMsg'
Learn More

5-methyl-2-phenyl-2H-1,2,3-triazole-4-carbonyl chloride, Maybridge

Brand:  Maybridge CC16202CB

Code : 31

Additional Details : CAS Number : 36401-55-5 Weight : 0.05025kg Transport : UN number : 3261 Chem class : 8 Pack group : II

Packaging Amber Glass Bottle
Quantity 250mg

 View more versions of this product

Product Code. 10426652

  • 28.00 € / 250mg


Estimated Shipment
29-03-2019
Add to basket

Specifications

Chemical Identifiers

36401-55-5
221.644
5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonyl chloride, 5-methyl-2-phenyl-1,2,3-triazole-4-carbonyl chloride, 2h-1,2,3-triazole-4-carbonylchloride, 5-methyl-2-phenyl, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbonyl chloride, 5-methyl-2-phenyl-4-triazolecarbonyl chloride, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbo nyl chloride, 5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonylchloride
5-methyl-2-phenyltriazole-4-carbonyl chloride
C10H8ClN3O
UJYBUZMRRLFXGM-UHFFFAOYSA-N
2776447
CC1=NN(N=C1C(=O)Cl)C2=CC=CC=C2

Specifications

5-Methyl-2-phenyl-2H-1, 2, 3-triazole-4-carbonyl chloride
250mg
97%
5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonyl chloride, 5-methyl-2-phenyl-1,2,3-triazole-4-carbonyl chloride, 2h-1,2,3-triazole-4-carbonylchloride, 5-methyl-2-phenyl, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbonyl chloride, 5-methyl-2-phenyl-4-triazolecarbonyl chloride, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbo nyl chloride, 5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonylchloride
CC1=NN(N=C1C(=O)Cl)C2=CC=CC=C2
221.644
221.65
Amber Glass Bottle
36401-55-5
C10H8ClN3O
UJYBUZMRRLFXGM-UHFFFAOYSA-N
5-methyl-2-phenyltriazole-4-carbonyl chloride
2776447
97%
Videos
SDS