missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiazole, 97%, ACROS Organics™

Brand:  Acros Organics 438940010

Code : 29

Additional Details : CAS Number : 3581-89-3 Weight : 0.05100kg Transport : UN number : 1993 Chem class : 3 Pack group : III


Quantity 1g

 View more versions of this product

Product Code. 11946781

  • 56.46 € / 1g

Estimated Shipment
22-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

3581-89-3
99.16
5-methylthiazole, thiazole, 5-methyl, 5-methyl-thiazole, 5-methyl thiazole, zlchem 1239, acmc-1cjhk, #, ksc222m5d
5-methyl-1,3-thiazole
C4H5NS
RLYUNPNLXMSXAX-UHFFFAOYSA-N
137980
CC1=CN=CS1

Specifications

5-Methylthiazole
Authentic
141°C to 142°C (993.0 hPa)
-40°C
3581-89-3
C4H5NS
RLYUNPNLXMSXAX-UHFFFAOYSA-N
5-methyl-1,3-thiazole
137980
Liquid
42°C
Glass Bottle
Colorless to Yellow
1g
96% min. (GC)
5-methylthiazole, thiazole, 5-methyl, 5-methyl-thiazole, 5-methyl thiazole, zlchem 1239, acmc-1cjhk, #, ksc222m5d
CC1=CN=CS1
99.16
99.16
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Methylthiazole

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS Documents