missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-1,10-phenanthroline, 98%, Thermo Scientific Chemicals

Catalog Number 11460273
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
4199-88-6
C12H7N3O2
225.207
MFCD00004981
PDDBTWXLNJNICS-UHFFFAOYSA-N
72790
5-nitro-1,10-phenanthroline
C1=CC2=CC(=C3C=CC=NC3=C2N=C1)[N+](=O)[O-]
This item is not returnable. View return policy
4199-88-6
C12H7N3O2
225.207
MFCD00004981
PDDBTWXLNJNICS-UHFFFAOYSA-N
72790
5-nitro-1,10-phenanthroline
C1=CC2=CC(=C3C=CC=NC3=C2N=C1)[N+](=O)[O-]

5-Nitro-1,10-phenanthroline complexes of iron find wide applications in redox titrations. It is used as a precursor for the synthesis of 5-amino-1,10-phenanthroline, 5-isothiocyanato-1,10-phenanthroline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitro-1,10-phenanthroline complexes of iron find wide applications in redox titrations. It is used as a precursor for the synthesis of 5-amino-1,10-phenanthroline, 5-isothiocyanato-1,10-phenanthroline.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 202°C to 204°C
Quantity 1 g
Beilstein 196245
Synonym 1,10-phenanthroline, 5-nitro, 5-nitro-1, 10-diazaphenanthrene, nitroferroin, 5-nitropyridino 3,2-h quinoline, acmc-209jmn, phhn01, 5nitro-1, 10-phenanthroline, 5-nitro-1,10-diazaphenanthrene, pddbtwxlnjnics-uhfffaoysa, 5-nitro-1,10-phenanthroline, crystalline
Solubility Information Soluble in ethanol. Insoluble in water.
Formula Weight 225.21
Percent Purity 98%
Chemical Name or Material 5-Nitro-1,10-phenanthroline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'