missing translation for 'onlineSavingsMsg'
Learn More

5-Nitroindole, 99%, Thermo Scientific Chemicals

Catalog Number p-173523
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6146-52-7
C8H6N2O2
162.15
MFCD00005673
OZFPSOBLQZPIAV-UHFFFAOYSA-N
5-nitroindole, 1h-indole, 5-nitro, indole, 5-nitro, unii-o2bhx6edbn, 5-nitro indole, ccris 3255, o2bhx6edbn, pubchem1711, acmc-209msr
22523
5-nitro-1H-indole
C1=CC2=C(C=CN2)C=C1[N+](=O)[O-]
This item is not returnable. View return policy
6146-52-7
C8H6N2O2
162.15
MFCD00005673
OZFPSOBLQZPIAV-UHFFFAOYSA-N
5-nitroindole, 1h-indole, 5-nitro, indole, 5-nitro, unii-o2bhx6edbn, 5-nitro indole, ccris 3255, o2bhx6edbn, pubchem1711, acmc-209msr
22523
5-nitro-1H-indole
C1=CC2=C(C=CN2)C=C1[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 140.0°C to 142.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 162.15
Percent Purity 99%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 5-Nitroindole, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Nitroindole
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'