Learn More
Invitrogen™ iBind™ Flex Replacement Well Inserts
Use with the iBind™ Flex Western Device (SLF2000). Invitrogen™ iBind™ Flex Multi Well Insert Replacement is a well insert that customizes the format to process up to six vertically cut strip blots. It features six wells on row 1 and 3.
Brand: Invitrogen™ SLF2006
Additional Details : Weight : 0.25000kg
Description
The iBind™ Flex Western System offers automated western blot processing. Using customized well inserts, it can accommodate various blot formats including midi, mini, and vertically cut strips. Based on Sequential Lateral Flow (SLF) technology, all blocking, antibody incubation and washing steps during immunodetection are hands-free. Compared to manual methods, the iBind Flex Western System requires less primary antibody, enables reproducible results and is compatible with downstream detection protocols.
Specifications
Multi-strip well insert with six wells on row 1 and 3 | |
Six wells on row 1 and 3 | |
Replacement Well Inserts | |
Western Blotting |
For Research Use Only. Not for use in diagnostic procedures.