missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 5 12-14 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
61.02 € - 237.00 €
Specifications
MWCO | 12 to 14 kDa |
---|
Includes:
- 1m or 5m of Spectra/Por 5 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
Product Code | Brand | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity & Availability | |||
15330782
|
Spectrum™
132757T |
62 mL/cm vol./length | 89 mm | 5 m Tubing/Kit |
237.00 €
5m |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15310782
|
Spectrum™
132754T |
18 mL/cm vol./length | 48 mm | 5 m Tubing/Kit |
195.00 €
5m |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15340782
|
Spectrum™
132 757T1 |
62 mL/cm vol./length | 89 mm | 1 m Tubing/Kit |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15320782
|
Spectrum™
132 754T1 |
18 mL/cm vol./length | 48 mm | 1 m Tubing/Kit |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- For reinforced for weight applications, or evaluating dialysis tubing for dialysis application
- Packaged dry without glycerin (no soaking required)
Specifications
12 to 14 kDa |