missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 5810 Centrifuges without Rotor

Brand:  Eppendorf™ 5810000017

UNSPSC : 41103903

Code : NEW

Additional Details : Weight : 85.00000kg

 View more versions of this product

Product Code. 10000461

  • 4478.00 € / Each


Call for latest local availability
01 885 5854
Add to basket

Specifications

Specifications

4 x 750mL
50 to 60Hz
345mm
<70dB(A)
535mm
230V, 50 to 60Hz, 900w
68Kg
200 to 14,000rpm
Centrifuge
20,913 x G
230V
68Kg
1 to 99min.
900W
5810
608mm
1Each
2 to 40°C
View accessory products
Alternative Products