missing translation for 'onlineSavingsMsg'
Learn More

5alpha-Pregnan-3beta,17alpha-diol-20-one, TRC

Product Code. 30011658
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011658 100 mg 100mg
30005258 250 mg 250mg
30005235 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011658 Supplier Toronto Research Chemicals Supplier No. TRCP711335100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 570-54-7
Molecular Formula C21 H34 O3
Molecular Weight (g/mol) 334.49
Synonym 3β,17-Dihydroxy-5α-pregnan-20-one;3β,17α-Dihydroxy-5α-pregnan-20-one;5α-Pregnane-3β,17α-diol-20-one;Allopregnane-3β,17α-diol-20-one;Reichstein's Substance L;Wintersteiner's Compound G;
IUPAC Name 1-[(3S,5S,8R,9S,10S,13S,14S,17R)-3,17-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-yl]ethanone
SMILES CC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CC[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H34O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h14-18,23-24H,4-12H2,1-3H3/t14-,15-,16+,17-,18-,19-,20-,21-/m0/s1
Formula Weight 334.2508
Chemical Name or Material 5alpha-Pregnan-3beta,17alpha-diol-20-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.