missing translation for 'onlineSavingsMsg'
Learn More

6,9-Diamino-2-ethoxyacridine lactate monohydrate, 95%, ACROS Organics™

Brand:  Acros Organics 406380050

Code : 29

Additional Details : CAS Number : 1837-57-6 Weight : 0.05500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10026023

  • 26.52 € / 5g

Estimated Shipment
28-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

6402-23-9
361.39
NYEPHMYJRNWPLA-UHFFFAOYSA-N
165457
CCOC1=CC2=C(C3=C(C=C(C=C3)N)N=C2C=C1)N.CC(C(=O)O)O.O
C3H6O3·H2O
MFCD00149646
ethacridine lactate monohydrate, ethodin, ethacridine lactate hydrate, ethacridine lactate salt, dsstox_cid_26274, dsstox_rid_81501, dsstox_gsid_46274, 7-ethoxyacridine-3,9-diamine; 2-hydroxypropanoic acid; hydrate, rivanol monohydrate
7-ethoxyacridine-3,9-diamine;2-hydroxypropanoic acid;hydrate

Specifications

6, 9-Diamino-2-ethoxyacridine lactate monohydrate
Glass bottle
243°C to 245°C
6402-23-9,1837-57-6
100.0
C3H6O3·H2O
15, 3771
Solubility in water: .. Other solubilities: slowly soluble in 15 parts water,soluble in 9 parts boiling water,soluble in 110 parts alcohol (22°C),soluble in 100 parts alcohol (20°C)
CCOC1=CC2=C(C3=C(C=C(C=C3)N)N=C2C=C1)N.CC(C(=O)O)O.O
361.39
361.39
95%
Authentic
Yellow
5g
94.0
95%
MFCD00149646
ethacridine lactate monohydrate, ethodin, ethacridine lactate hydrate, ethacridine lactate salt, dsstox_cid_26274, dsstox_rid_81501, dsstox_gsid_46274, 7-ethoxyacridine-3,9-diamine; 2-hydroxypropanoic acid; hydrate, rivanol monohydrate
NYEPHMYJRNWPLA-UHFFFAOYSA-N
7-ethoxyacridine-3,9-diamine;2-hydroxypropanoic acid;hydrate
165457
Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 6,9-Diamino-2-ethoxyacridine lactate monohydrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents