Learn More
6-Benzyloxypyridine-3-boronic acid, 97%, ACROS Organics™
Brand: Acros Organics 445320010
Code : 29
Additional Details : CAS Number : 929250-35-1 Weight : 0.05100kg
Quantity | 1g |
---|
Chemical Identifiers
929250-35-1 | |
229.04 | |
2-benzyloxypyridine-5-boronic acid, 6-benzyloxypyridine-3-boronic acid, 6-benzyloxy pyridin-3-yl boronic acid, 6-benzyloxy pyridine-3-boronic acid, 6-benzyloxy pyridin-3-ylboronic acid, 2-benzyloxy-pyridine-5-boronic acid, 6-benzyloxy pyridin-3-yl-3-boronic acid, 2-benzyloxypyridin-5-yl boronic acid, 2-benzyloxy pyridine-5-ylboronic acid | |
(6-phenylmethoxypyridin-3-yl)boronic acid |
C12H12BNO3 | |
CUZOGAUUSKBDAO-UHFFFAOYSA-N | |
17750216 | |
B(C1=CN=C(C=C1)OCC2=CC=CC=C2)(O)O |
Specifications
6-Benzyloxypyridine-3-boronic acid | |
107°C to 110°C | |
929250-35-1 | |
C12H12BNO3 | |
CUZOGAUUSKBDAO-UHFFFAOYSA-N | |
(6-phenylmethoxypyridin-3-yl)boronic acid | |
17750216 | |
97% |
Glass Bottle | |
1g | |
96% min. (HPLC) | |
2-benzyloxypyridine-5-boronic acid, 6-benzyloxypyridine-3-boronic acid, 6-benzyloxy pyridin-3-yl boronic acid, 6-benzyloxy pyridine-3-boronic acid, 6-benzyloxy pyridin-3-ylboronic acid, 2-benzyloxy-pyridine-5-boronic acid, 6-benzyloxy pyridin-3-yl-3-boronic acid, 2-benzyloxypyridin-5-yl boronic acid, 2-benzyloxy pyridine-5-ylboronic acid | |
B(C1=CN=C(C=C1)OCC2=CC=CC=C2)(O)O | |
229.04 | |
229.04 |
Safety and Handling
- 6-Benzyloxypyridine-3-boronic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.