missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1,4-benzodioxane, 97%, ACROS Organics™

Brand:  Acros Organics 441700050

Code : 29

Additional Details : CAS Number : 52287-51-1 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11936211

  • 37.37 € / 5g

Estimated Shipment: 06-07-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

52287-51-1
215.05
6-bromo-1,4-benzodioxane, 6-bromo-2,3-dihydrobenzo b 1,4 dioxine, 3,4-ethylenedioxybromobenzene, 6-bromo-2,3-dihydro-1,4-benzodioxin, 1,4-benzodioxin, 6-bromo-2,3-dihydro, 4-bromo-1,2-ethylenedioxybenzene, 3,4-ethylendioxybromobenzene, 6-bromo-2,3-dihydro-benzo 1,4 dioxine, pubchem3736, acmc-209kyk
6-bromo-2,3-dihydro-1,4-benzodioxine
C8H7BrO2
LFCURAJBHDNUNG-UHFFFAOYSA-N
104141
C1COC2=C(O1)C=CC(=C2)Br

Specifications

6-Bromo-1, 4-benzodioxane
>110°C
Glass Bottle
1.598
Colorless to Yellow
52287-51-1
100.0
C8H7BrO2
LFCURAJBHDNUNG-UHFFFAOYSA-N
6-bromo-2,3-dihydro-1,4-benzodioxine
104141
Liquid
1.5980g/mL
Authentic
1.588
259°C to 260°C
5g
96.0
96% min. (GC)
6-bromo-1,4-benzodioxane, 6-bromo-2,3-dihydrobenzo b 1,4 dioxine, 3,4-ethylenedioxybromobenzene, 6-bromo-2,3-dihydro-1,4-benzodioxin, 1,4-benzodioxin, 6-bromo-2,3-dihydro, 4-bromo-1,2-ethylenedioxybenzene, 3,4-ethylendioxybromobenzene, 6-bromo-2,3-dihydro-benzo 1,4 dioxine, pubchem3736, acmc-209kyk
C1COC2=C(O1)C=CC(=C2)Br
215.05
215.05
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 6-Bromo-1,4-benzodioxane

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates