missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Chloropurine riboside, 98%

Brand:  Alfa Aesar™ H59634.06

Code : NEW

Additional Details : CAS Number : 5399-87-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15476018

  • 466.00 € / 5g


Estimated Shipment date
from Supplier 23-05-2019
Add to basket

Description

Description

Applications
6-Chloropurine riboside is used to study the kinetics and substrate specificity of adenosine deaminase. 6-Chloropurine riboside is benzoylated to facilitate synthesis of nucleoside derivatives such as 9-(2,3-Di-deoxy-2-fluoro-β-D-threo-pentofuranosyl)adenine. It , especially after phosphorylation to NMP, NDP or NTP, is used as a purine substrate analogue in studies with enzymes such as Inosine monophosphate dehydrogenase (IMPDH); bacteriophage T4 RNA-ligase (EC 6.5.1.3) and pancreatic fibonuclease A.

Solubility
Soluble in water. (10.6 mg/mL)

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

5399-87-1
286.672
XHRJGHCQQPETRH-KQYNXXCUSA-N
93003
C1=NC2=C(C(=N1)Cl)N=CN2C3C(C(C(O3)CO)O)O
C10H11ClN4O4
MFCD00005738
6-chloropurine riboside, chloropurine riboside, 2r,3r,4s,5r-2-6-chloro-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 6-chloropurine ribonucleoside, 6-chloropurine-9-riboside, 6-chloro-9-ribofuranosyl-9h-purine, 9h-purine, 6-chloro-9-ribofuranosyl, 6-chloropurineriboside, 6-chloro-9-beta-d-ribofuranosyl-9h-purine, 6-chloronebularine
(2R,3R,4S,5R)-2-(6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol

Specifications

6-Chloropurine riboside
158°C to 162°C (decomposition)
5399-87-1
MFCD00005738
6-chloropurine riboside, chloropurine riboside, 2r,3r,4s,5r-2-6-chloro-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 6-chloropurine ribonucleoside, 6-chloropurine-9-riboside, 6-chloro-9-ribofuranosyl-9h-purine, 9h-purine, 6-chloro-9-ribofuranosyl, 6-chloropurineriboside, 6-chloro-9-beta-d-ribofuranosyl-9h-purine, 6-chloronebularine
C1=NC2=C(C(=N1)Cl)N=CN2C3C(C(C(O3)CO)O)O
286.672
286.67
98%
White
5g
C10H11ClN4O4
40573
XHRJGHCQQPETRH-KQYNXXCUSA-N
(2R,3R,4S,5R)-2-(6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
93003
Powder
Videos
SDS