Learn More
6-Chloropurine riboside, 98%, Thermo Scientific Chemicals
Description
6-Chloropurine riboside is used to study the kinetics and substrate specificity of adenosine deaminase. 6-Chloropurine riboside is benzoylated to facilitate synthesis of nucleoside derivatives such as 9-(2,3-Di-deoxy-2-fluoro-β-D-threo-pentofuranosyl)adenine. It , especially after phosphorylation to NMP, NDP or NTP, is used as a purine substrate analogue in studies with enzymes such as Inosine monophosphate dehydrogenase (IMPDH); bacteriophage T4 RNA-ligase (EC 6.5.1.3) and pancreatic fibonuclease A.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
6-Chloropurine riboside is used to study the kinetics and substrate specificity of adenosine deaminase. 6-Chloropurine riboside is benzoylated to facilitate synthesis of nucleoside derivatives such as 9-(2,3-Di-deoxy-2-fluoro-β-D-threo-pentofuranosyl)adenine. It , especially after phosphorylation to NMP, NDP or NTP, is used as a purine substrate analogue in studies with enzymes such as Inosine monophosphate dehydrogenase (IMPDH); bacteriophage T4 RNA-ligase (EC 6.5.1.3) and pancreatic fibonuclease A.
Solubility
Soluble in water. (10.6 mg/mL)
Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 158°C to 162°C (decomposition) |
Color | White |
Quantity | 1 g |
Beilstein | 40573 |
Solubility Information | Soluble in water. (10.6mg/mL) |
Formula Weight | 286.67 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | 6-Chloropurine riboside |
RUO – Research Use Only