missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-1-indanone, 97%, ACROS Organics™

Brand:  Acros Organics 442330010

Code : 29

Additional Details : CAS Number : 1481-32-9 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 11934861

  • 69.72 € / 1g

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1481-32-9
150.15
LVUUCFIQQHEFEJ-UHFFFAOYSA-N
1519464
FC1=CC2=C(CCC2=O)C=C1
C9H7FO
MFCD01318147
6-fluoro-1-indanone, 6-fluoro-2,3-dihydro-1h-inden-1-one, 6-fluoro-indan-1-one, 6-fluoroindan-1-one, 6-fluroro-1-indanone, 1h-inden-1-one, 6-fluoro-2,3-dihydro, 6-fluoroindanone, 6fluoro-1-indanone, pubchem8856, acmc-1bwfy
6-fluoro-2,3-dihydroinden-1-one

Specifications

6-Fluoro-1-indanone
Glass Bottle
58°C to 61°C
1481-32-9
100.0
C9H7FO
6-fluoro-1-indanone, 6-fluoro-2,3-dihydro-1h-inden-1-one, 6-fluoro-indan-1-one, 6-fluoroindan-1-one, 6-fluroro-1-indanone, 1h-inden-1-one, 6-fluoro-2,3-dihydro, 6-fluoroindanone, 6fluoro-1-indanone, pubchem8856, acmc-1bwfy
FC1=CC2=C(CCC2=O)C=C1
150.15
150.15
97%
Authentic
White to Brown
1g
96.0
96% min. (GC)
MFCD01318147
LVUUCFIQQHEFEJ-UHFFFAOYSA-N
6-fluoro-2,3-dihydroinden-1-one
1519464
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 6-Fluoro-1-indanone

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents