missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 6-Methyl-2-(2-thienyl)benzimidazole, 97%

Brand:  Alfa Aesar™ H58782.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15474988

  • 328.00 € / 1g

Estimated Shipment date
from Supplier 24-07-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

3878-24-8
214.29
QIYVOSKFSQXPCG-UHFFFAOYSA-N
924806
C12H10N2S
MFCD14618840
5-methyl-2-thiophen-2-yl-1h-benzoimidazole, 5-methyl-2-2-thienyl benzimidazole, 6-methyl-2-2-thienyl benzimidazole, 2-5-methylbenzimidazol-2-yl thiophene, 5-methyl-2-thien-2-yl-1h-benzimidazole, 2-2-thienyl-5-methyl-1h-benzimidazole, 5-methyl-2-2-thienyl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzo d imidazole, 6-methyl-2-thiophen-2-yl-1h-benzo d imidazole

Specifications

6-Methyl-2-(2-thienyl)benzimidazole
1g
C12H10N2S
5-methyl-2-thiophen-2-yl-1h-benzoimidazole, 5-methyl-2-2-thienyl benzimidazole, 6-methyl-2-2-thienyl benzimidazole, 2-5-methylbenzimidazol-2-yl thiophene, 5-methyl-2-thien-2-yl-1h-benzimidazole, 2-2-thienyl-5-methyl-1h-benzimidazole, 5-methyl-2-2-thienyl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzimidazole, 5-methyl-2-thiophen-2-yl-1h-benzo d imidazole, 6-methyl-2-thiophen-2-yl-1h-benzo d imidazole
214.29
214.28
97%
Brown to Yellow
3878-24-8
MFCD14618840
QIYVOSKFSQXPCG-UHFFFAOYSA-N
924806
Solid
Videos
SDS