Learn More
6-piperidin-1-ylnicotinaldehyde, Maybridge™
Brand: Maybridge MO07199DE
Code : 31
Additional Details : CAS Number : 241816-11-5 Weight : 0.05500kg
Quantity | 5g |
---|
Chemical Identifiers
241816-11-5 | |
190.246 | |
6-piperidin-1-ylnicotinaldehyde, 6-piperidin-1-yl nicotinaldehyde, 6-piperidin-1-yl pyridine-3-carbaldehyde, 6-piperidinonicotinaldehyde, 6-1-piperidinyl nicotinaldehyde, 6-1-piperidinyl pyridine-3-carboxaldehyde, 6-piperidylpyridine-3-carbaldehyde, acmc-20alxp, 3-pyridinecarboxaldehyde,6-1-piperidinyl, 3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-5'-carbaldehyde | |
6-piperidin-1-ylpyridine-3-carbaldehyde |
C11H14N2O | |
RKIMIISNVCLRLA-UHFFFAOYSA-N | |
2763980 | |
C1CCN(CC1)C2=NC=C(C=C2)C=O |
Specifications
6-Piperidin-1-ylnicotinaldehyde | |
5g | |
97% | |
6-piperidin-1-ylnicotinaldehyde, 6-piperidin-1-yl nicotinaldehyde, 6-piperidin-1-yl pyridine-3-carbaldehyde, 6-piperidinonicotinaldehyde, 6-1-piperidinyl nicotinaldehyde, 6-1-piperidinyl pyridine-3-carboxaldehyde, 6-piperidylpyridine-3-carbaldehyde, acmc-20alxp, 3-pyridinecarboxaldehyde,6-1-piperidinyl, 3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-5'-carbaldehyde | |
C1CCN(CC1)C2=NC=C(C=C2)C=O | |
190.246 | |
190.24 |
Amber glass bottle | |
241816-11-5 | |
C11H14N2O | |
RKIMIISNVCLRLA-UHFFFAOYSA-N | |
6-piperidin-1-ylpyridine-3-carbaldehyde | |
2763980 | |
97% |