missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-(tert-Butyl) 3-ethyl 2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 326.41 |
Percent Purity | 95% |
Chemical Name or Material | 6-(tert-Butyl) 3-ethyl 2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'