missing translation for 'onlineSavingsMsg'
Learn More

LMS™ Series 4 Model 600 Cooled Incubator, 600 L, Stainless Steel

Temperature Range: -10°C to +50°C

Brand:  LMS™ 600


Code : 04

Additional Details : Weight : 1.00000kg

Product Code. 11877602

  • 16801.00 € / Each
Estimated Shipment: 01-07-2024
to see stock.
Add to basket

Description

Description

  • Reliable and accurate temperature control
  • Temperature range -10.0°C to +50.0°C
  • Models within this series include 600L and 1200L capacity incubators as either single or dual temperature (24 hour or 7 day cycle) versions, or a fully programmable 1200L capacity incubator providing multiple temperature control
  • Stainless steel exterior and interior construction with foamed polyurethane insulation
  • Energy efficient and eco-friendly
  • P.I.D. heating and cooling to ensure stable internal temperature
  • Pre-set safety cut-out
  • Hermetically sealed refrigeration system
  • Door lock
  • Controls easy to read and simple to use
  • Temperature variation with average workload ±0.5°C
  • Full range of factory-fitted optional extras available, including LED lighting, access porthole, inner perspex doors, outer glazed doors, alarm system, internal socket and chart recorder
  • Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests
Specifications

Specifications

ISO 9001:2008
Freestanding incubator
Stainless Steel
550 mm x 530 mm
Stainless Steel
-10°C to +50°C
Digital
600
CFC-free
±0.5°C
29.5 in.
750 mm
81.9 in.
2080 mm
26.4 in.
160 kg
230/240 V
Standard digital
600 L
Fan assisted air circulation via premixing chamber
Optional LED
5
Refrigerated Incubator
Foamed polyurethane
1
±0.1°C
2 Years
23.6 in.
600 mm
59.0 in.
1500 mm
670 mm
240 V, 50 Hz
50 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers