Learn More
Hamilton™ 600 Microliter Syringes, No Needle Included
The 600 series syringes are a half stroke version of the original 700 series syringes. This series of syringes is ideal for dispensing volumes from 0.25 µL up to 5 µL. The 600 series is often used in applications that require one handed operation because of the short stroke length and the reinforced plunger.
172.00 €
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11522505
|
Hamilton™
7632-01 |
2.5 μL |
172.00 €
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11532505
|
Hamilton™
7633-01 |
5 μL |
172.00 €
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Half stroke version of the original icroliter 700 series syringes
- Ideal for dispensing volumes from 0.25 μL up to 5 μL
- Often used in applications that require one handed operation because of the short stroke length and the reinforced plunger
Specifications
22s G |