Learn More
7-Bromo-2,3-dioxoindoline, 97%, Maybridge™
Brand: Maybridge MO07293DA
Code : 31
Additional Details : CAS Number : 20780-74-9 Weight : 0.05100kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
20780-74-9 | |
226.029 | |
OCVKSIWBTJCXPV-UHFFFAOYSA-N | |
2302353 | |
C1=CC2=C(C(=C1)Br)NC(=O)C2=O |
C8H4BrNO2 | |
MFCD00774354 | |
7-bromoisatin, 7-bromoindoline-2,3-dione, 7-bromo-2,3-dioxoindoline, 1h-indole-2,3-dione, 7-bromo, 7-bromoindole-2,3-dione, 7-bromo-2,3-dihydro-1h-indole-2,3-dione, 7-bromoindole-1h-2,3-dione, 7-bromo-2,3-indolinedione, 7-bromo isatin, 7-bromo-isatin | |
7-bromo-1H-indole-2,3-dione |
Specifications
7-Bromo-2, 3-dioxoindoline | |
1g | |
97.0 | |
C8H4BrNO2 | |
7-bromoisatin, 7-bromoindoline-2,3-dione, 7-bromo-2,3-dioxoindoline, 1h-indole-2,3-dione, 7-bromo, 7-bromoindole-2,3-dione, 7-bromo-2,3-dihydro-1h-indole-2,3-dione, 7-bromoindole-1h-2,3-dione, 7-bromo-2,3-indolinedione, 7-bromo isatin, 7-bromo-isatin | |
C1=CC2=C(C(=C1)Br)NC(=O)C2=O | |
226.029 | |
226.03 |
Amber Glass Bottle | |
20780-74-9 | |
97% | |
MFCD00774354 | |
OCVKSIWBTJCXPV-UHFFFAOYSA-N | |
7-bromo-1H-indole-2,3-dione | |
2302353 | |
97% |