missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-6-nitro-4(3H)-quinazolinone, 97%, Thermo Scientific™

Catalog Number 15264854
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
53449-14-2
C8H4ClN3O3
225.59
URDYTQYZXZKBQT-UHFFFAOYSA-N
7-chloro-6-nitroquinazolin-4 3h-one, 7-chloro-4-hydroxy-6-nitroquinazoline, 7-chloro-6-nitro-4-hydroxyquinazoline, 6-nitro-7-chloro-4-hydroxyquinazoline, 7-chloro-6-nitroquinazolin-4-ol, 7-chloro-6-nitroquinazolin-4-one, 7-chloro-6-nitro-quinazolin-4-one, 7-chloro-6-nitro-3h-quinazolin-4-one, 4 1h-quinazolinone, 7-chloro-6-nitro, 7-chloro-6-nitro-3,4-dihydroquinazolin-4-one
12441237
7-chloro-6-nitro-1H-quinazolin-4-one
C1=C2C(=CC(=C1[N+](=O)[O-])Cl)NC=NC2=O
This item is not returnable. View return policy
53449-14-2
C8H4ClN3O3
225.59
URDYTQYZXZKBQT-UHFFFAOYSA-N
7-chloro-6-nitroquinazolin-4 3h-one, 7-chloro-4-hydroxy-6-nitroquinazoline, 7-chloro-6-nitro-4-hydroxyquinazoline, 6-nitro-7-chloro-4-hydroxyquinazoline, 7-chloro-6-nitroquinazolin-4-ol, 7-chloro-6-nitroquinazolin-4-one, 7-chloro-6-nitro-quinazolin-4-one, 7-chloro-6-nitro-3h-quinazolin-4-one, 4 1h-quinazolinone, 7-chloro-6-nitro, 7-chloro-6-nitro-3,4-dihydroquinazolin-4-one
12441237
7-chloro-6-nitro-1H-quinazolin-4-one
C1=C2C(=CC(=C1[N+](=O)[O-])Cl)NC=NC2=O
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: .. Other solubilities: slightly soluble in dmf
Formula Weight 225.59
Percent Purity 97%
Chemical Name or Material 7-Chloro-6-nitro-4(3H)-quinazolinone
compliance-icons
Product Identifier
  • 7-Chloro-6-nitroquinazolin-4-ol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'