missing translation for 'onlineSavingsMsg'
Learn More

7-Nitroindole-2-carboxylic acid, 90%, Thermo Scientific™

Catalog Number 10004962
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
6960-45-8
C9H6N2O4
206.157
BIUCOFQROHIAEO-UHFFFAOYSA-N
7-nitro-1h-indole-2-carboxylic acid, 7-nitroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 7-nitro, 7-nitroindole-2-carboxylicacid, dna base excision repair pathway inhibitor, maybridge1_004786, indole-2-carboxylic acid, 7-nitro, pubchem9857, acmc-1b8pb, 1h-indole-2-carboxylicacid, 7-nitro
81409
C1=CC2=C(C(=C1)[N+](=O)[O-])NC(=C2)C(=O)O
This item is not returnable. View return policy
6960-45-8
C9H6N2O4
206.157
BIUCOFQROHIAEO-UHFFFAOYSA-N
7-nitro-1h-indole-2-carboxylic acid, 7-nitroindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 7-nitro, 7-nitroindole-2-carboxylicacid, dna base excision repair pathway inhibitor, maybridge1_004786, indole-2-carboxylic acid, 7-nitro, pubchem9857, acmc-1b8pb, 1h-indole-2-carboxylicacid, 7-nitro
81409
C1=CC2=C(C(=C1)[N+](=O)[O-])NC(=C2)C(=O)O
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 206.16
Percent Purity 90%
Chemical Name or Material 7-Nitro-1H-indole-2-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'