Learn More
Cytiva Grade 740E Filter Discs
Meets FDA/WHO antibiotic-assay control disc standards. Whatman™ Grade 740E Filter Discs are thick, highly absorbent paper discs made of high-purity cotton linter for use in antibiotic-assay procedures.
Brand: Cytiva 10328170
Code : A0
Additional Details : Weight : 0.07200kg
Description
These filter discs can be used to determine the type and level of microorganisms and to check their sensitivity to antibiotics and chemotherapeutic agents.
- Specified in standard tests for antibiotic susceptibility as well as antibiotic levels in milk
- Features coarse porosity with particle retention of 20-30μm
- Enables a fast flow/filtration rate
Order Info
dimGrade
Specifications
Cotton | |
Fast | |
740E | |
Filter Disc | |
12.7 mm |
20 to 30 μm | |
Antibiotic-assay procedures | |
1000Pack | |
1/2 in. |