missing translation for 'onlineSavingsMsg'
Learn More

8-Amino-5-bromoisoquinoline, 97%, ACROS Organics™

Brand:  Acros Organics 431080050

Code : 29

Additional Details : CAS Number : 90721-35-0 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11966501

  • 131.00 € / 5g

In stock, ready to ship: 1

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

90721-35-0
223.07
8-amino-5-bromoisoquinoline, 5-bromoisoquinolin-8-ylamine, 5-bromo-8-isoquinolineamine, 5-bromo-isoquinolin-8-ylamine, 8-isoquinolinamine, 5-bromo, 5-bromo-8-isoquinolinamine, 8-amino-5-bromisoquinoline, pubchem20332, 5-bromo-8-aminoisoquinoline
5-bromoisoquinolin-8-amine
C9H7BrN2
BTFXYVXBWMRHIB-UHFFFAOYSA-N
16218302
C1=CC(=C2C=CN=CC2=C1N)Br

Specifications

8-Amino-5-bromoisoquinoline
White to Brown
5g
96.0
96% min. (HPLC)
8-amino-5-bromoisoquinoline, 5-bromoisoquinolin-8-ylamine, 5-bromo-8-isoquinolineamine, 5-bromo-isoquinolin-8-ylamine, 8-isoquinolinamine, 5-bromo, 5-bromo-8-isoquinolinamine, 8-amino-5-bromisoquinoline, pubchem20332, 5-bromo-8-aminoisoquinoline
C1=CC(=C2C=CN=CC2=C1N)Br
223.07
223.07
97%
Authentic
187.4°C to 189.4°C
90721-35-0
100.0
C9H7BrN2
BTFXYVXBWMRHIB-UHFFFAOYSA-N
5-bromoisoquinolin-8-amine
16218302
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 8-Amino-5-bromoisoquinoline, 97%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS Documents