missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-1-octanol, 95%, Thermo Scientific Chemicals

Catalog Number p-7077412
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
50816-19-8
C8H17BrO
209.13
GMXIEASXPUEOTG-UHFFFAOYSA-N
8-bromo-1-octanol, 1-octanol, 8-bromo, 8-bromooctanol, octamethylene bromohydrin, 8-bromo octanol, 1-bromooctan-8-ol, 8-bromo-1-actanol, 8-bromooctane-1-ol, 1-bromo-octan-8-ol, 8-bromo-octan-1-ol
162607
8-bromooctan-1-ol
C(CCCCBr)CCCO
This item is not returnable. View return policy
50816-19-8
C8H17BrO
209.13
GMXIEASXPUEOTG-UHFFFAOYSA-N
8-bromo-1-octanol, 1-octanol, 8-bromo, 8-bromooctanol, octamethylene bromohydrin, 8-bromo octanol, 1-bromooctan-8-ol, 8-bromo-1-actanol, 8-bromooctane-1-ol, 1-bromo-octan-8-ol, 8-bromo-octan-1-ol
162607
8-bromooctan-1-ol
C(CCCCBr)CCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.2200g/mL
Boiling Point 79.0°C to 80.0°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.22
Formula Weight 209.13
Percent Purity 95%
Chemical Name or Material 8-Bromo-1-octanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'