missing translation for 'onlineSavingsMsg'
Learn More

8-(chloromethyl)-6-fluoro-4H-1,3-benzodioxine, 97%, Maybridge™

Brand:  Maybridge CD08167EA

Code : 31

Additional Details : CAS Number : 131728-94-4 Weight : 0.06000kg Transport : UN number : 3261 Chem class : 8 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 10002464

  • 190.13 € / 10g

Estimated Shipment: 02-09-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

131728-94-4
202.61
FMONGDHUPLQOCP-UHFFFAOYSA-N
2780137
FC1=CC(CCl)=C2OCOCC2=C1
C9H8ClFO2
MFCD00052603
8-chloromethyl-6-fluoro-4h-1,3-benzodioxine, 8-chloromethyl-6-fluoro-4h-benzo d 1,3 dioxine, 4h-1,3-benzodioxin,8-chloromethyl-6-fluoro, 8-chloromethyl-6-fluoro-2,4-dihydro-1,3-benzodioxine, 8-chloromethyl-6-fluorobenzo-1,3-dioxane, acmc-1c5x1, 8-chloromethyl-6-fluoro-4h-benzo 1,3 dioxine, 8-chloromethyl-6-fluoro-2h,4h-1,3-benzodioxine, 8-chloromethyl-6-fluoro-2h,4h-benzo e 1,3-dioxin
8-(chloromethyl)-6-fluoro-4H-1,3-benzodioxine

Specifications

8-(Chloromethyl)-6-fluoro-4H-1, 3-benzodioxine
131728-94-4
100.0
C9H8ClFO2
8-chloromethyl-6-fluoro-4h-1,3-benzodioxine, 8-chloromethyl-6-fluoro-4h-benzo d 1,3 dioxine, 4h-1,3-benzodioxin,8-chloromethyl-6-fluoro, 8-chloromethyl-6-fluoro-2,4-dihydro-1,3-benzodioxine, 8-chloromethyl-6-fluorobenzo-1,3-dioxane, acmc-1c5x1, 8-chloromethyl-6-fluoro-4h-benzo 1,3 dioxine, 8-chloromethyl-6-fluoro-2h,4h-1,3-benzodioxine, 8-chloromethyl-6-fluoro-2h,4h-benzo e 1,3-dioxin
FC1=CC(CCl)=C2OCOCC2=C1
202.61
202.61
10g
96.5
97%
MFCD00052603
FMONGDHUPLQOCP-UHFFFAOYSA-N
8-(chloromethyl)-6-fluoro-4H-1,3-benzodioxine
2780137
97%
Videos
SDS Documents
Certificates