missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ 900 mm ReVo™ Class II, Type A2 Microbiological Safety Cabinet
Shop All Labconco Corporation ProductsDescription
- Provide personnel, product and environmental protection from biohazardous agents and hazardous particulates.
- Other appropriate applications include work with antineoplastic drugs, genetic material, and additional substances that generate hazardous airborne particulates..
- All models are EN 12469 compliant with additional certification by TÜV Nord.
- These biological safety cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection.
- When canopy-connected, the biosafety cabinets may be used for applications involving minute quantities of volatile toxic chemicals and tracer amounts of radionuclides as an adjunct to microbiological research.
- During operation, room air is drawn into the inlet grill.
- The air in the plenum beneath the work surface is a mixture of unfiltered room air and air that has just passed through the work area.
- This contaminated air is drawn by the blower through the back plenum of the cabinet, where approximately 70% of the air is recirculated through the supply HEPA filter and back over the work area.
- The balance of the contaminated air is discharged to the environment after passing through the exhaust HEPA filter.
Specifications
Specifications
| Certifications/Compliance | EN 12469 Certified by TÜV |
| Chamber Material | Stainless Steel (304) |
| Product Type | Class II Biosafety Cabinet |
| Noise Level | 43 dBA |
| Outlets | 2 |
| Power Consumption | 85 W |
| Sash Opening (Metric) | 200 mm |
| Exhaust Air Filter | HEPA Exhaust Filter |
| Filter Type | H14 HEPA |
| Includes | Base Stand Not Included |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?