missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex 95282 Medium Tune-Up Insole For Uvex 1 and 2 - Width 11
Tune-Up antistatic insole for reducing fatigue and supporting arch
21.80 €
Specifications
Color | Black, White |
---|---|
For Use With (Equipment) | Uvex 1 and 2 |
Material | Microvelour |
Disposable | No |
Gender | Unisex |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17298011
|
Uvex
9528235 |
35 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17208021
|
Uvex
9528236 |
36 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17218021
|
Uvex
9528237 |
37 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17228021
|
Uvex
9528238 |
38 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17238021
|
Uvex
9528239 |
39 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17248021
|
Uvex
9528240 |
40 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17258021
|
Uvex
9528241 |
41 |
21.80 €
1 pair |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
17268021
|
Uvex
9528242 |
42 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17278021
|
Uvex
9528243 |
43 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17288021
|
Uvex
9528244 |
44 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17298021
|
Uvex
9528245 |
45 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17208031
|
Uvex
9528246 |
46 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17218031
|
Uvex
9528247 |
47 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17228031
|
Uvex
9528248 |
48 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17238031
|
Uvex
9528249 |
49 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17248031
|
Uvex
9528250 |
50 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17258031
|
Uvex
9528251 |
51 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17268031
|
Uvex
9528252 |
52 |
21.80 €
1 pair |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
Uvex Tune-Up Comfortable Medium Insole 95282
- Antistatic insole concept for use in safety footwear
- Designed to reduce fatigue by increasing wearer comfort and enhancing ergonomics by optimum support of arch of feet
- Good cushioning
- Relief of pressure marks
- Non-irritating, comfortable surface structure
- Breathable and absorbs moisture
- Antibacterial
- Antistatic
- Useable for ESD-footwear
- Uvex 1, Uvex 2, Uvex 1 Duo, Uvex Xenova™ Light
- Article No. 95282
- Standard EN ISO 20345:2011 S1 SRC
- Sizes: 35 to 52
Specifications
Black, White | |
Microvelour | |
Unisex | |
Yes |
Uvex 1 and 2 | |
No | |
Standard width | |
No |