Learn More
Greiner Bio-One Non-treated 96-well Microplates
Manufactured without surface treatment to create low biomolecular binding for sensitive biochemical screening applications. Greiner Bio One™ Non-treated 96-Well Assay Microplates are available in polystyrene or polypropylene with a range of well designs.
Brand:  Greiner Bio-One 655101
Additional Details : Weight : 3.15000kg
 View more versions of this product
Product Code. 10252711
  • 120.00 € / Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
  • Footprint compatible with automated systems
  • Can be traced to production through defined LOT number system
  • Free of detectable endotoxins (0.003 EU/mL) and regularly tested using an FDA-approved kinetic turbidimetric LAL-test (Limulus Amoebocyte Assay)
  • Manufactured from raw materials tested for leachables
  • Free of detectable DNase, RNase and human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Models with chimney design allow each well to stand on its owndiminishing the risk of sample carryover and cross contamination
Specifications
Specifications
Alphanumeric
Standard
8 x 12
Polystyrene
Non-sterile
382 μL
Yes
-20 to +60°C
DIN ISO 9001
Without Lid
Clear
96
Non-treated
Flat Bottom (F-well)
100/Cs.