missing translation for 'onlineSavingsMsg'
Learn More

Norharman, 98%, Thermo Scientific Chemicals

Catalog Number p-2703632
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
244-63-3
C11H8N2
168.20
MFCD00004956
AIFRHYZBTHREPW-UHFFFAOYSA-N
9h-pyrido 3,4-b indole, norharman, norharmane, beta-carboline, carbazoline, 2,9-diazafluorene, 9h-beta-carboline, 2-azacarbazole, .beta.-carboline
64961
CHEBI:109895
9H-pyrido[3,4-b]indole
N1C2=C(C=CC=C2)C2=C1C=NC=C2
This item is not returnable. View return policy
244-63-3
C11H8N2
168.20
MFCD00004956
AIFRHYZBTHREPW-UHFFFAOYSA-N
9h-pyrido 3,4-b indole, norharman, norharmane, beta-carboline, carbazoline, 2,9-diazafluorene, 9h-beta-carboline, 2-azacarbazole, .beta.-carboline
64961
CHEBI:109895
9H-pyrido[3,4-b]indole
N1C2=C(C=CC=C2)C2=C1C=NC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 199.0°C to 201.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Tertiary base)
Packaging Glass bottle
Quantity 100 mg
Beilstein 23, II, 224
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 168.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Norharman
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'