Learn More
Thermo Scientific™ PCR Plate, 384-well, standard, red
Thermo Scientific™ ABgene 384-Well PCR Plate Full Skirted PCR Plates are thin walled for efficient heat transfer and are compatible with most major thermal cyclers.
Brand: Thermo Scientific™ AB1384R
Code : A0
Additional Details : Weight : 0.80000kg
Description
Thermo Scientific™ ABgene™ 384-Well Full Skirted PCR Plates are thin walled for efficient heat transfer and compatible with most major thermal cyclers.
- Raised rim design around each well enables secure sealing and safeguards against evaporation
- Uniform, thin well walls deliver maximum and consistent heat transfer for equally high performance from every sample
- SBS footprint
- 40μL maximum well volume, 25μL working volume
- Cut Corner: A24
Industry-leading Manufacturing and QC Testing for ultimate product reliability:
- Cleanroom Produced—To avoid particulate and biological contaminants that can interfere with molecular biology applications
- Virgin Medical Grade Polypropylene—Specialized polymer is highly inert and will not interfere with PCR reaction components
- Integrity Testing—Every well of every plate is visually inspected and tested using an electrostatic pinhole detection method
- PCR Functional Assays—Products are tested in PCR assays to verify clean negative controls and consistent technical replicates
- Evaporation Testing—Samples from each lot are run through PCR cycling to test sealing performance
- Biological Testing—All lots are biologically tested to certify them free of RNase, DNase, and DNA
Format Options:
Bar Coded Plates
- Feature two industry standard 128 bar codes carefully designed and positioned for compatibility with all major bar code readers
- Codes are random and each bar code also includes a human readable format
- Wide temperature tolerance (-196° to +120°C)
- Proprietary coating for superior scratch-resistance
- Precise sizing and placement for reliable scanning
- Custom bar coding also available, please enquire
White Plates
- Designed specifically for qPCR
- Reflect significantly more signal than traditional clear plates, which ensures that even the lowest levels of fluorescence are detected
- Enable consistent signal reflection—the opaque well walls prevent signal from passing through to the cycler block where it can be inconsistently reflected or absorbed, keeping variations in the cycler block from affecting your qPCR data
Colored Plates
- Great for sample organization and tracking
- Available in five distinct colors: Blue, Green, Purple, Red, and Yellow
Compatible with:
Sealing Options: Adhesive Sealing, Heat Sealing.
Recommended for:
PCR; qPCR / Real-Time PCR
Specifications
No Barcode | |
50 Plate | |
PCR Microplate | |
Skirted | |
40 μL | |
25 μL |
Certified RNase/DNase-free | |
Red | |
Real-Time PCR Systems or Thermal Cyclers | |
Polypropylene | |
384 | |
Standard |